Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synpo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191963.1 | NP_001178892.1 | 3 - 4 | 1171 | 100 | |
XM_008761470.1 | XP_008759692.1 | 4 - 5 | 1178 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |