Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100992.2 | NP_001094462.2 | 1 - 2 | 213 | 65 | |
XM_006232728.2 | XP_006232790.1 | 1 - 2 | 476 | 65 | ||
GenBank mRNA | BC127525.1 | - | 1 - 2 | 213 | 7114610 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |