Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271054.1 | NP_001257983.1 | 4 - 5 | 530 | 118 | |
GenBank mRNA | AF441249.1 | - | 3 - 4 | 307 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |