Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031064.2 | NP_112326.1 | 7 - 8 | 699 | 64 | |
GenBank mRNA | BC058495.1 | - | 7 - 8 | 703 | 6891104 | 64 |
FQ215131.1 | - | 4 - 5 | 372 | 64 | ||
M22756.1 | - | 7 - 8 | 624 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |