Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033359.3 | NP_203523.2 | 1 - 2 | 500 | 83 | |
XM_008764864.1 | XP_008763086.1 | 1 - 2 | 1181 | 83 | ||
GenBank mRNA | AB056655.1 | - | 1 - 2 | 72 | 83 | |
AF333245.1 | - | 1 - 2 | 91 | 83 | ||
AF334379.1 | - | 1 - 2 | 91 | 83 | ||
AY066001.1 | - | 1 - 2 | 300 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |