Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039505.2 | NP_001034594.1 | 1 - 2 | 518 | 88 | |
XM_008758689.1 | XP_008756911.1 | 1 - 2 | 575 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |