Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134858.3 | NP_001128330.1 | 1 - 2 | 1033 | 115 | |
XM_006229337.2 | XP_006229399.1 | 1 - 2 | 1028 | 115 | ||
GenBank mRNA | AB091769.1 | - | 1 - 2 | 941 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |