Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pate-f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267780.1 | NP_001254709.1 | 2 - 3 | 195 | 70 | |
GenBank mRNA | JF412806.1 | - | 2 - 3 | 195 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Probe spans exons |