Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MNN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178363.1 | NP_009571.1 | 1 | 1349 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |