Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178537.3 | NP_009748.3 | 1 | 250 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Inventoried Both primers and probe map within a single exon |