Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178564.3 | NP_009775.3 | 1 | 1750 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |