Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001179494.1 | NP_012120.1 | 1 | 1455 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |