Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001182689.1 | NP_013908.1 | 1 | 656 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |