Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001182950.3 | NP_014287.3 | 1 - 2 | 1275 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Saccharomyces cerevisiae Probe spans exons |