Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190203.1 | NP_001177132.1 | 3 - 4 | 207 | 85 | |
XM_005656224.3 | XP_005656281.1 | 300 | 85 | |||
GenBank mRNA | AY610382.1 | - | 3 - 4 | 172 | 85 | |
DY428718.1 | - | 1 - 2 | 108 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |