Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005157.1 | NP_001005157.1 | 5 - 6 | 730 | 81 | |
XM_005665792.3 | XP_005665849.1 | 966 | 81 | |||
XM_005665793.3 | XP_005665850.1 | 953 | 81 | |||
XM_005665794.3 | XP_005665851.1 | 1017 | 81 | |||
XM_021098341.1 | XP_020954000.1 | 1397 | 81 | |||
GenBank mRNA | AK236289.1 | - | 6 - 7 | 904 | 81 | |
AK238677.1 | - | 6 - 7 | 919 | 81 | ||
AK389641.1 | - | 6 - 7 | 1014 | 81 | ||
AK389706.1 | - | 6 - 7 | 921 | 81 | ||
AK399598.1 | - | 6 - 7 | 921 | 81 | ||
EU282340.1 | - | 5 - 6 | 1381 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |