Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100736733 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001315796.1 | NP_001302725.1 | 5 - 6 | 721 | 100 | |
XM_021088677.1 | XP_020944336.1 | 878 | 100 | |||
GenBank mRNA | AK231223.1 | - | 6 - 7 | 865 | 100 | |
AK392957.1 | - | 6 - 7 | 867 | 100 | ||
AY609994.1 | - | 6 - 7 | 851 | 100 | ||
EU560428.1 | - | 5 - 6 | 721 | 100 |
Gene Symbol: LOC110255228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021089549.1 | XP_020945208.1 | 100 | |||
XM_021089550.1 | XP_020945209.1 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |