Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KISS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134964.1 | NP_001128436.1 | 1 - 2 | 131 | 53 | |
XM_005656644.2 | XP_005656701.1 | 672 | 53 | |||
GenBank mRNA | AB466320.1 | - | 2 - 3 | 404 | 53 | |
EU586514.1 | - | 1 - 2 | 131 | 53 | ||
EU934235.1 | - | 1 - 2 | 131 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons |