Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHSG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005652369.3 | XP_005652426.1 | 1198 | 94 | ||
GenBank mRNA | AK233033.1 | - | 5 - 6 | 754 | 94 | |
X56021.1 | - | 668 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |