Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044546.3 | NP_001038011.2 | 4 | 550 | 120 | |
GenBank mRNA | AK239299.1 | - | 4 | 549 | 120 | |
AY902753.1 | - | 4 | 550 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Amplicon spans exons and probe does not span exons |