Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048070.1 | NP_001041535.1 | 5 | 451 | 121 | |
GenBank mRNA | AK234891.1 | - | 5 | 461 | 121 | |
AK399738.1 | - | 5 | 472 | 121 | ||
AY609569.1 | - | 5 | 457 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |