Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP3A39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214422.1 | NP_999587.1 | 13 | 1545 | 84 | |
GenBank mRNA | AF109068.1 | - | 13 | 1545 | 84 | |
KP687262.1 | - | 14 | 1553 | 84 |
Gene Symbol: CYP3A22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021085547.1 | XP_020941206.1 | 1816 | 84 | ||
GenBank mRNA | AK399802.1 | - | 14 | 1835 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon |