Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112679.1 | NP_001106149.1 | 3 | 444 | 81 | |
GenBank mRNA | EF577504.1 | - | 3 | 444 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |