Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097439.1 | NP_001090908.1 | 8 - 9 | 1329 | 128 | |
GenBank mRNA | AK391731.1 | - | 3 - 4 | 273 | 128 | |
AK392225.1 | - | 8 - 9 | 1483 | 128 | ||
AM042539.1 | - | 8 - 9 | 1329 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |