Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100193.1 | NP_001093663.1 | 4 - 5 | 388 | 71 | |
GenBank mRNA | DQ351286.1 | - | 4 - 5 | 388 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |