Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101033.1 | NP_001094503.1 | 5 - 6 | 620 | 103 | |
XM_005672829.3 | XP_005672886.1 | 2119 | 103 | |||
XM_021077174.1 | XP_020932833.1 | 2119 | 103 | |||
GenBank mRNA | AK237864.1 | - | 5 - 6 | 626 | 103 | |
AK350647.1 | - | 5 - 6 | 637 | 103 | ||
DQ859019.1 | - | 5 - 6 | 620 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |