Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DXO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101826.1 | NP_001095296.1 | 6 - 7 | 1243 | 59 | |
GenBank mRNA | AK351293.1 | - | 6 - 7 | 1405 | 59 | |
EU282361.1 | - | 5 - 6 | 1131 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |