Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110427.2 | NP_001103897.2 | 2 - 3 | 151 | 119 | |
NM_001257090.1 | NP_001244019.1 | 2 - 3 | 206 | 119 | ||
GenBank mRNA | AB193117.1 | - | 2 - 3 | 206 | 119 | |
AB193119.1 | - | 2 - 3 | 151 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Probe spans exons |