Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110432.1 | NP_001103902.1 | 5 - 6 | 575 | 68 | |
XM_005673032.3 | XP_005673089.1 | 562 | 68 | |||
GenBank mRNA | AK233642.1 | - | 5 - 6 | 525 | 68 | |
AK235189.1 | - | 5 - 6 | 548 | 68 | ||
AK348007.1 | - | 5 - 6 | 501 | 68 | ||
AK396192.1 | - | 5 - 6 | 547 | 68 | ||
AK397035.1 | - | 5 - 6 | 551 | 68 | ||
AK397163.1 | - | 5 - 6 | 550 | 68 | ||
AK399170.1 | - | 5 - 6 | 550 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |