Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114289.2 | NP_001107761.2 | 1 - 2 | 103 | 75 | |
GenBank mRNA | AK352281.1 | - | 1 - 2 | 135 | 75 | |
EU364897.2 | - | 1 - 2 | 103 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Inventoried Probe spans exons |