Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123134.1 | NP_001116606.1 | 5 - 6 | 671 | 124 | |
XM_005671253.3 | XP_005671310.1 | 888 | 124 | |||
XM_013983513.2 | XP_013838967.1 | 717 | 124 | |||
XM_021071782.1 | XP_020927441.1 | 894 | 124 | |||
GenBank mRNA | AK230645.1 | - | 6 - 7 | 698 | 124 | |
AK389355.1 | - | 6 - 7 | 750 | 124 | ||
EU442570.1 | - | 5 - 6 | 671 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |