Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123134.1 | NP_001116606.1 | 8 - 9 | 972 | 74 | |
XM_005671253.3 | XP_005671310.1 | 1189 | 74 | |||
XM_013983513.2 | XP_013838967.1 | 1018 | 74 | |||
XM_021071782.1 | XP_020927441.1 | 1195 | 74 | |||
GenBank mRNA | AK230645.1 | - | 9 - 10 | 999 | 74 | |
AK389355.1 | - | 9 - 10 | 1051 | 74 | ||
EU442570.1 | - | 8 - 9 | 972 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |