Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNXB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123204.1 | NP_001116676.1 | 10774 | 92 | ||
XM_021098327.1 | XP_020953986.1 | 13564 | 92 | |||
XM_021098328.1 | XP_020953987.1 | 13510 | 92 | |||
XM_021098329.1 | XP_020953988.1 | 13510 | 92 | |||
XM_021098330.1 | XP_020953989.1 | 13267 | 92 | |||
XM_021098331.1 | XP_020953990.1 | 13267 | 92 | |||
XM_021098332.1 | XP_020953991.1 | 13237 | 92 | |||
XM_021098333.1 | XP_020953992.1 | 13210 | 92 | |||
XM_021098334.1 | XP_020953993.1 | 12802 | 92 | |||
XM_021098335.1 | XP_020953994.1 | 12616 | 92 | |||
XM_021098336.1 | XP_020953995.1 | 12616 | 92 | |||
XM_021098337.1 | XP_020953996.1 | 12520 | 92 | |||
XM_021098338.1 | XP_020953997.1 | 11542 | 92 | |||
XM_021098339.1 | XP_020953998.1 | 7474 | 92 | |||
XM_021098340.1 | XP_020953999.1 | 12505 | 92 | |||
GenBank mRNA | EU275239.1 | - | 2 - 3 | 569 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |