Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAPLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004028.1 | NP_001004028.1 | 3 - 4 | 794 | 126 | |
XM_005661538.3 | XP_005661595.1 | 1105 | 126 | |||
XM_005661539.3 | XP_005661596.1 | 1025 | 126 | |||
XM_013994961.2 | XP_013850415.1 | 1053 | 126 | |||
XM_013994962.2 | XP_013850416.1 | 1138 | 126 | |||
GenBank mRNA | AK350792.1 | - | 4 - 5 | 866 | 126 | |
Y00165.1 | - | 794 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Probe spans exons |