Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213816.1 | NP_998981.1 | 4 - 5 | 964 | 66 | |
GenBank mRNA | AF156712.1 | - | 4 - 5 | 964 | 66 | |
AK240579.1 | - | 2 - 3 | 578 | 66 | ||
AK349681.1 | - | 4 - 5 | 1091 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |