Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214034.1 | NP_999199.1 | 4 | 1450 | 97 | |
GenBank mRNA | X04413.1 | - | 1450 | 97 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |