Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214064.1 | NP_999229.1 | 9 | 1553 | 104 | |
XM_005656645.3 | XP_005656702.1 | 1770 | 104 | |||
GenBank mRNA | AK232750.1 | - | 9 | 1576 | 104 | |
AK233513.1 | - | 9 | 1565 | 104 | ||
AK393950.1 | - | 9 | 1579 | 104 | ||
KC734480.1 | - | 8 | 1499 | 104 | ||
M32031.1 | - | 9 | 1553 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon |