Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214118.2 | NP_999283.1 | 2 - 3 | 242 | 91 | |
GenBank mRNA | AY116504.1 | - | 2 - 3 | 243 | 91 | |
D21074.1 | - | 2 - 3 | 207 | 91 | ||
DQ108393.1 | - | 2 - 3 | 207 | 91 | ||
U61139.1 | - | 2 - 3 | 207 | 91 | ||
U67175.1 | - | 2 - 3 | 207 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |