Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU1F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214163.1 | NP_999328.1 | 1 - 2 | 265 | 65 | |
XM_005657130.3 | XP_005657187.1 | 143 | ||||
GenBank mRNA | AF016251.2 | - | 1 - 2 | 265 | 65 | |
AF016382.1 | - | 1 - 2 | 265 | 65 | ||
HM163575.1 | - | 1 - 2 | 154 | 65 | ||
Y12475.1 | - | 130 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |