Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214175.1 | NP_999340.1 | 2 - 3 | 161 | 120 | |
XM_005659588.3 | XP_005659645.1 | 270 | 120 | |||
XM_005659589.3 | XP_005659646.1 | 465 | 120 | |||
XM_021082942.1 | XP_020938601.1 | 141 | 120 | |||
GenBank mRNA | AJ005981.1 | - | 2 - 3 | 161 | 120 | |
AK232838.1 | - | 2 - 3 | 311 | 120 | ||
AK239939.1 | - | 2 - 3 | 297 | 120 | ||
AK395513.1 | - | 3 - 4 | 218 | 120 | ||
AY610426.1 | - | 2 - 3 | 200 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Probe spans exons |