Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214200.2 | NP_999365.1 | 7 - 8 | 989 | 79 | |
XM_005660074.3 | XP_005660131.1 | 79 | ||||
XM_005660075.3 | XP_005660132.1 | 79 | ||||
XM_013993524.2 | XP_013848978.1 | 79 | ||||
XM_021082975.1 | XP_020938634.1 | 79 | ||||
GenBank mRNA | AB527057.1 | - | 6 - 7 | 79 | ||
AF234676.2 | - | 7 - 8 | 990 | 79 | ||
AK232656.1 | - | 7 - 8 | 79 | |||
AY550037.1 | - | 7 - 8 | 987 | 79 | ||
DQ066659.1 | - | 6 - 7 | 79 | |||
KC733245.1 | - | 6 - 7 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |