Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214206.1 | NP_999371.1 | 2 - 3 | 219 | 70 | |
GenBank mRNA | AK232208.1 | - | 2 - 3 | 226 | 70 | |
AK233316.1 | - | 2 - 3 | 235 | 70 | ||
AK345472.1 | - | 2 - 3 | 215 | 70 | ||
AK345483.1 | - | 2 - 3 | 462 | 70 | ||
AK391018.1 | - | 2 - 3 | 246 | 70 | ||
AK400768.1 | - | 2 - 3 | 246 | 70 | ||
AY609777.1 | - | 2 - 3 | 317 | 70 | ||
U62253.1 | - | 2 - 3 | 219 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |