Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPYR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214243.1 | NP_999408.1 | 1 | 1352 | 67 | |
GenBank mRNA | AB021678.1 | - | 1 | 1352 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |