Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214286.1 | NP_999451.1 | 7 - 8 | 1296 | 94 | |
XM_021072174.1 | XP_020927833.1 | 1349 | 94 | |||
GenBank mRNA | AK344311.1 | - | 7 - 8 | 1335 | 94 | |
AY559452.1 | - | 7 - 8 | 1275 | 94 | ||
AY559453.1 | - | 7 - 8 | 1275 | 94 | ||
AY559454.1 | - | 7 - 8 | 1275 | 94 | ||
AY686760.1 | - | 7 - 8 | 1275 | 94 | ||
AY686761.1 | - | 7 - 8 | 1275 | 94 | ||
HQ699490.1 | - | 7 - 8 | 1193 | 94 | ||
X62984.1 | - | 1313 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |