Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214307.1 | NP_999472.1 | 2 - 3 | 224 | 98 | |
GenBank mRNA | AK343797.1 | - | 2 - 3 | 224 | 98 | |
AK344811.1 | - | 2 - 3 | 234 | 98 | ||
AK399705.1 | - | 2 - 3 | 259 | 98 | ||
AY271355.1 | - | 2 - 3 | 224 | 98 | ||
AY610247.1 | - | 2 - 3 | 193 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Probe spans exons |