Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL13RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214341.1 | NP_999506.1 | 4 - 5 | 478 | 128 | |
GenBank mRNA | AK395683.1 | - | 4 - 5 | 523 | 128 | |
AY266142.1 | - | 4 - 5 | 478 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |