Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214446.1 | NP_999611.1 | 2 - 3 | 292 | 75 | |
XM_005659277.3 | XP_005659334.1 | 456 | 75 | |||
XM_021083511.1 | XP_020939170.1 | 520 | 75 | |||
GenBank mRNA | AK349136.1 | - | 3 - 4 | 390 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |