Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185133.2 | NP_001172062.1 | 2 - 3 | 158 | 97 | |
GenBank mRNA | AK343813.1 | - | 2 - 3 | 158 | 97 | |
AK392192.1 | - | 2 - 3 | 166 | 97 | ||
AY609839.1 | - | 2 - 3 | 155 | 97 | ||
FJ487630.1 | - | 2 - 3 | 159 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |