Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163649.2 | NP_001157121.1 | 1 - 2 | 442 | 97 | |
XM_005656620.3 | XP_005656677.2 | 601 | 97 | |||
XM_005656621.3 | XP_005656678.2 | 661 | 97 | |||
GenBank mRNA | AK234261.1 | - | 2 - 3 | 501 | 97 | |
AK349885.1 | - | 2 - 3 | 575 | 97 | ||
AK393012.1 | - | 2 - 3 | 498 | 97 | ||
GQ245762.1 | - | 1 - 2 | 444 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |