Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZPBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164513.1 | NP_001157985.1 | 6 - 7 | 796 | 109 | |
XM_005653912.3 | XP_005653969.1 | 109 | ||||
XM_013979545.2 | XP_013834999.1 | 109 | ||||
GenBank mRNA | GQ369764.1 | - | 7 - 8 | 109 | ||
GQ369765.1 | - | 6 - 7 | 796 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |